Learn More
Human CXCR6 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CXCR6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1010910 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | O00574 |
| Gene Alias | Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.