Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR6 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB699100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB699100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB699100 Supplier R&D Systems Supplier No. MAB699100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR6 Monoclonal specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry, CyTOF-reported.

Specifications

Antigen CXCR6
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 56811
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00574
Gene Alias Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186
Gene Symbols CXCR6
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CXCR6 Met1-Leu342 Accession # O00574
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10663
Test Specificity Stains human CXCR6 transfectants but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.