Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR6 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 14 publications
$186.30 - $456.00
Specifications
Antigen | CXCR6 |
---|---|
Clone | 56811 |
Dilution | Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB699P025
![]() |
R&D Systems
FAB699P025 |
25 Tests |
Each for $186.30
|
|
|||||
FAB699P100
![]() |
R&D Systems
FAB699P100 |
100 Tests |
Each for $456.00
|
|
|||||
Description
CXCR6 Monoclonal specifically detects CXCR6 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR6 | |
Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
Human | |
Bonzo, BONZOG-protein coupled receptor bonzo, CD186, CD186 antigen, CDw186, chemokine (C-X-C motif) receptor 6, C-X-C chemokine receptor type 6, CXCR6, CXC-R6, CXCR-6, G protein-coupled receptor, STRL33, STRL33G-protein coupled receptor STRL33, TYMSTRCD186 | |
CXCR6 | |
IgG2b | |
Protein A or G purified from ascites | |
Stains human CXCR6 transfectants but not irrelevant transfectants. |
56811 | |
Flow Cytometry | |
R-PE | |
Mouse | |
O00574 | |
10663 | |
NS0 mouse myeloma cell line transfected with human CXCR6 Met1-Leu342 Accession # O00574 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title