Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR7/RDC-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB42273G100UG
This item is not returnable.
View return policy
Description
CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for Flow Cytometry.Specifications
CXCR7/RDC-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P25106 | |
CXCR7 | |
Human CXCR7 encoding plasmid Accession # P25106 | |
RUO | |
57007 | |
Human | |
IgG1 |
Flow Cytometry | |
11G8 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
Mouse | |
100 μg | |
Primary | |
Detects human CXCR7/RDC-1 in flow cytometry of five distinct human CXCR7 transfectants, but not their respective parental lines. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction