Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CXCR7/RDC-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB42273RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB42273RSP 25 μg
MA42273R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB42273RSP Supplier R&D Systems Supplier No. MAB42273RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen CXCR7/RDC-1
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 11G8R
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/mL, Immunohistochemistry 5-25 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P25106
Gene Alias ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Host Species Mouse
Immunogen Human CXCR7 encoding plasmid Accession # P25106
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 57007
Test Specificity Detects human CXCR7/RDC-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.