Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CXCR7/RDC-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | CXCR7/RDC-1 |
---|---|
Clone | 358440 |
Dilution | Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB42274SP
![]() |
R&D Systems
MAB42274SP |
25 μg |
Each for $200.50
|
|
|||||
MAB42274100
![]() |
R&D Systems
MAB42274100 |
100 μg |
Each for $468.00
|
|
|||||
Description
CXCR7/RDC-1 Monoclonal antibody specifically detects CXCR7/RDC-1 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
CXCR7/RDC-1 | |
Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
NS0 mouse myeloma cell line transfected with human CXCR7/RDC-1, Met1-Lys362, Accession # AAA62370 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
358440 | |
Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
AAA62370 | |
57007 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title