Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cyclin B1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC60001T100UG
This item is not returnable.
View return policy
Description
Cyclin B1 Monoclonal specifically detects Cyclin B1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Cyclin B1 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CCNB, CCNB1, cyclin B1, G2/mitotic-specific cyclin B1, G2/mitotic-specific cyclin-B1 | |
E. coli-derived recombinant human Cyclin B1 Met1-Pro91 Accession # P14635 | |
RUO | |
891 | |
Human | |
IgG |
Flow Cytometry | |
2061D | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P14635 | |
Rabbit | |
100 μg | |
Primary | |
Detects human Cyclin B1 in direct ELISAs and Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction