Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cyclin D1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4314SP
This item is not returnable.
View return policy
Description
Cyclin D1 Monoclonal specifically detects Cyclin D1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
Cyclin D1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P24385 | |
CCND1 | |
E. coli-derived recombinant human Cyclin D1 Met1-Ile295 Accession # P24385 | |
25 μg | |
Primary | |
Detects human Cyclin D1 in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry, Immunocytochemistry | |
328303 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, CCND1, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), Cyl-1, G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
595 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction