Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cyclin E1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6810SP
This item is not returnable.
View return policy
Description
Cyclin E1 Polyclonal specifically detects Cyclin E1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Cyclin E1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCNE, CCNE1, CCNEcyclin Es, cyclin E1, cyclin Et, G1/S-specific cyclin-E1 | |
Sheep | |
Affinity Purified | |
RUO | |
898 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL | |
P24864 | |
CCNE1 | |
E. coli-derived recombinant human Cyclin E1 Met16-Lys143 Accession # P24864 | |
25 μg | |
Primary | |
Detects human Cyclin E1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction