Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Cynomolgus Monkey FCAR/CD89 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3939
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3939 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3939 Supplier R&D Systems Supplier No. MAB3939
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen FCAR/CD89
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 488032
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P24071
Gene Alias CD89, CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Gene Symbols FCAR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FCAR/CD89 Gln22-Lys287 Accession # P24071
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2204
Test Specificity Detects human FCAR/CD89 in direct ELISAs. In direct ELISAs, no cross-reactivity with Fc gamma RIA, RIIA, or RIIIB is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.