Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CYP7A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$544.12
Specifications
| Antigen | CYP7A1 |
|---|---|
| Clone | 946902 |
| Dilution | Immunocytochemistry 1-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9120100
![]() |
R&D Systems
MAB9120100 |
100 μg |
Each for $544.12
|
|
|||||
|
MAB9120SP
|
R&D Systems
MAB9120SP |
25 μg | N/A | N/A | N/A | ||||
Description
CYP7A1 Monoclonal specifically detects CYP7A1 in Human samples. It is validated for Immunocytochemistry.Specifications
| CYP7A1 | |
| Immunocytochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cholesterol 7-alpha-hydroxylase, cholesterol 7-alpha-monooxygenase, CP7A, CYP7, CYP7MGC126826, CYPVII, Cytochrome P450 7A1, cytochrome P450, family 7, subfamily A, polypeptide 1, cytochrome P450, subfamily VIIA (cholesterol 7 alpha-monooxygenase), cytochrome P450, subfamily VIIA polypeptide 1, EC 1.14.13, EC 1.14.13.17, MGC138389, polypeptide 1 | |
| CYP7A1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 946902 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P22680 | |
| 1581 | |
| Human CYP7A1 synthetic peptide Accession # P22680 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CYP7A1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title