Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cystatin C Antibody, R&D Systems™

Mouse Monoclonal Antibody
$214.00 - $561.50
Specifications
Antigen | CystatinC |
---|---|
Clone | 197820R |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8μg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11962RSP
![]() |
R&D Systems
MAB11962RSP |
25 μg |
Each for $214.00
|
|
|||||
AB11962R100
![]() |
R&D Systems
MAB11962R100 |
100 μg |
Each for $561.50
|
|
|||||
Description
Cystatin C Monoclonal specifically detects Cystatin C in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
CystatinC | |
ELISA Capture (Matched Antibody Pair) 2-8μg/mL | |
Monoclonal | |
Supernatant | |
Human | |
P01034 | |
1471 | |
IgG2b | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
197820R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
ARMD11, bA218C14.4 (cystatin C), CST3, cystatin 3, cystatin C, cystatin C (amyloid angiopathy and cerebral hemorrhage), cystatin-3, cystatin-C, Gamma-trace, MGC117328, Neuroendocrine basic polypeptide, Post-gamma-globulin | |
Mouse myeloma cell line NS0-derived recombinant human Cystatin C Ser27-Ala146 Accession # P01034 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human Cystatin C in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title