Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cystatin E/M Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1286
This item is not returnable.
View return policy
Description
Cystatin E/M/CST6 Polyclonal specifically detects Cystatin E/M/CST6 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cystatin E/M/CST6 | |
Polyclonal | |
Biotin | |
Q15828 | |
CST6 | |
Mouse myeloma cell line NS0-derived recombinant human Cystatin E/M Arg29-Met149 Accession # Q15828 | |
50 μg | |
Primary | |
Detects human Cystatin E/M in Western blots. In Western blots, approximately 20% cross-reactivity with recombinant mouse Cystatin E/M is observed, less than 5% cross-reactivity with recombinant human (rh) Cystatin D is observed, and less than 1% cross-reactivity with rhCystatin A, rhCystatin B, rhCystatin C, rhCystatin S, rhCystatin SA, rhCystatin SN, rhFetuin A, and rhFetuin B is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
CST6, cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor | |
Goat | |
Affinity Purified | |
RUO | |
1474 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction