Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cystatin F Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $410.00
Specifications
Antigen | Cystatin F |
---|---|
Clone | 292103 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Cystatin F Monoclonal specifically detects Cystatin F in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cystatin F | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
O76096 | |
8530 | |
Mouse myeloma cell line NS0-derived recombinant human Cystatin F Gly20-His145 Accession # O76096 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Cystatin F in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Cystatin A, B, C, D, E/M, S, recombinant mouse (rm) Cystatin/Stefin Homolog, rhFetuin A, rhFetuin B, rhHPRG, rhKininogen, or rhKininostatin is observed. |
292103 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CMAP, CST7, Cystatin 7, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
CST7 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title