Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cystatin S Antibody, R&D Systems™

Goat Polyclonal Antibody
$176.44 - $532.55
Specifications
| Antigen | Cystatin S |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
Description
Cystatin S Polyclonal specifically detects Cystatin S in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cystatin S | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CST4, cystatin 4, cystatin S, Cystatin-4, cystatin-S, cystatin-SA-III, MGC71923, Salivary acidic protein 1 | |
| CST4 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| P01036 | |
| 1472 | |
| Mouse myeloma cell line NS0-derived recombinant human Cystatin S Ser21-Ala141 Accession # P01036 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Cystatin S in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human (rh) Cystatin SA and rhCystatin SN is observed and 5% cross-reactivity with rhCystatin C and rhCystatin D is observed, and less than 1% cross-reactivity with rhCystatin A, rhCystatin B, rhCystatin E/M, rhFetuin A, and rhFetuin B is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title