Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Cytochrome c Quantikine ELISA Kit

Catalog No. p-4753575
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human Cytochrome c Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Cytochrome c Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Cytochrome c in cell lysates (20 ul), subcellular fractions (100 ul).

Specifications

Assay Range 0.63 - 20ng/mL
Assay Sensitivity 0.31ng/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Lysates, Subcellular Fractions
Specificity Natural Human Cytochrome C
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 54205
Quantity 1 Plate
Sample Volume Cell Lysates (20uL),Subcellular Fractions (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CYCHCS, CYCS, cytochrome c, cytochrome c, somatic, THC4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.