Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cytohesin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4807SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4807SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4807SP Supplier R&D Systems Supplier No. AF4807SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Cytohesin-1 Polyclonal specifically detects Cytohesin-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cytohesin-1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q15438
Gene Alias B2-1, CYTH1, cytoadhesin 1, cytohesin 1, cytohesin-1, Cytohesin1, D17S811ECYTOHESIN-1, FLJ34050, FLJ41900, homolog of secretory protein SEC7, PH, SEC7 and coiled-coil domain-containing protein 1, pleckstrin homology, Sec7 and coiled-coil domains 1, PSCD1, SEC7, SEC7 homolog B2-1
Gene Symbols CYTH1
Host Species Goat
Immunogen E. coli-derived recombinant human Cytohesin-1 Met1-Tyr102 Accession # Q15438
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9267
Test Specificity Detects endogenous human Cytohesin-1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.