Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cytokeratin 18 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM7619
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM7619 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM7619 Supplier R&D Systems Supplier No. BAM7619
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytokeratin 18 Monoclonal specifically detects Cytokeratin 18 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Cytokeratin 18
Applications Immunocytochemistry
Classification Monoclonal
Clone 810811
Concentration LYOPH
Conjugate Biotin
Dilution Immunocytochemistry 25 ug/mL
Gene Accession No. P05783
Gene Alias Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, Keratin 18, keratin, type I cytoskeletal 18, keratin-18, KRT18
Gene Symbols KRT18
Host Species Mouse
Immunogen E. coli-derived recombinant human Cytokeratin 18 Ala239-Asp397 Accession # P05783
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) 3875
Test Specificity Cytokeratin 18 antibodies are ideal for immunocytochemistry colocalization studies in intermediate filaments. The unconjugated antibody detects human Cytokeratin 18 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.