Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cytokeratin 18/KRT18 ELISA Kit

Catalog No. p-7219324
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

Catalog No. EH145RBX5

Supplier: Invitrogen™ EH145RBX5

Only null left
Add to Cart
Add to Cart

ELISA

Human CytokeRatin 18/KRT18 quantitates human Cytokeratin 18/KRT18 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cytokeratin 18/KRT18.

Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number cb83; Cell proliferation-inducing gene 46 protein; CK 18; CK18; CK-18; CYK18; cytokeratin 18; cytokeratin endo B; cytokeratin type -1; DAPK-1; Cytokeratin18; cytokeratin-18; dapk1; Dorsal aorta proneprin kinesin-1; DreK18; Endo B; I79
Assay Range 0.164 to 40 ng/mL
Assay Sensitivity 0.164 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3875
Gene Symbol KRT18
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Cytokeratin 18
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.