Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cytokeratin 19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $567.90
Specifications
| Antigen | Cytokeratin 19 |
|---|---|
| Clone | 963420 |
| Dilution | Western Blot 0.25 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL, Dual RNAscope ISH-IHC |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH) |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB35061SP
![]() |
R&D Systems
MAB35061SP |
25 μg |
Each for $188.28
|
|
|||||
MAB35061100
![]() |
R&D Systems
MAB35061100 |
100 μg |
Each for $567.90
|
|
|||||
Description
Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, Dual RNAscope ISH-IHC.Specifications
| Cytokeratin 19 | |
| Western Blot 0.25 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL, Dual RNAscope ISH-IHC | |
| Monoclonal | |
| Mouse | |
| Human | |
| P08727 | |
| 3880 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 963420 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH) | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, EndoC, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, Krt19, MGC15366 | |
| E.coli-derived recombinant human Cytokeratin 19 Gln311-Lys370 Accession # P08727 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cytokeratin 19 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title