Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cytokeratin 19 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Cytokeratin 19 |
---|---|
Clone | 1042608 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB35063SP
![]() |
R&D Systems
MAB35063SP |
25 μg |
Each for $200.50
|
|
|||||
MAB35063100
![]() |
R&D Systems
MAB35063100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Cytokeratin 19 Monoclonal antibody specifically detects Cytokeratin 19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, ImmunocytochemistrySpecifications
Cytokeratin 19 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
40-kDa keratin intermediate filament, CK19, CK-19, cytokeratin 19, EndoC, K19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, Keratin-19, Krt19, MGC15366 | |
E. coli-derived human Cytokeratin 19, Gln311-Lys370, Accession # P08727 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1042608 | |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P08727 | |
3880 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title