Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human Cytokeratin 19/KRT19 ELISA Kit

Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Description
Human CytokeRatin 19/KRT19 quantitates human Cytokeratin 19/KRT19 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cytokeratin 19/KRT19.
Cytokeratin 19 is part of a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight from 40-68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. For example, cytokeratin 19 is not expressed in hepatocytes, therefore, it is useful in the identification of liver metastasis.

Specifications
Specifications
Accession Number | 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; KRT19; MGC15366; Type I keratin Ka19 |
Assay Range | 0.061 to 15 ng/mL |
Assay Sensitivity | 0.061 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Sample Type | Plasma, Serum, Supernatant |
For Use With (Equipment) | Colorimetric Microplate Reader |
Gene ID (Entrez) | 3880 |
Gene Symbol | KRT19 |
Interassay CV | <12% |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction