Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cytosolic beta-Glucosidase/GBA3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5969SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5969SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5969SP Supplier R&D Systems Supplier No. MAB5969SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cytosolic beta-Glucosidase/GBA3
Applications Western Blot
Classification Monoclonal
Clone 728714
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9H227
Gene Alias CBG, CBGL1, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic betaGlucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, GBA3, GLUC, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878
Gene Symbols GBA3
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Cytosolic beta-Glucosidase/GBA3 Thr13-Leu469 Accession # Q9H227
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57733
Test Specificity Detects human Cytosolic beta-Glucosidase/GBA3 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.