Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cytosolic Sulfotransferase 1B1/ SULT1B1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5959
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5959 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5959 Supplier R&D Systems Supplier No. AF5959
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Cytosolic Sulfotransferase 1B1/SULT1B1 Polyclonal specifically detects Cytosolic Sulfotransferase 1B1/SULT1B1 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Cytosolic Sulfotransferase 1B1/SULT1B1
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 110 ug/mL
Gene Accession No. O43704
Gene Alias EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.4, MGC13356, ST1B1, ST1B2, ST1B2sulfotransferase family cytosolic 1B member 1, Sulfotransferase 1B1, Sulfotransferase 1B2, sulfotransferase family, cytosolic, 1B, member 1, SULT1B1, SULT1B2, Thyroid hormone sulfotransferase
Gene Symbols SULT1B1
Host Species Sheep
Immunogen E. coli-derived recombinant human Cytosolic Sulfotransferase 1B1/SULT1B1 Leu2-Ile296 Accession # O43704
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27284
Test Specificity Detects human Cytosolic Sulfotransferase 1B1/SULT1B1 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) SULT1A1 and rhSULT1A3 is observed and less than 2% cross-reactivity with rhSULT2A1, rhSULT4A1, rhSULT2B1, and rhSULT1E1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.