Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cytosolic Sulfotransferase 1C2/SULT1C2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7458
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7458 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7458 Supplier R&D Systems Supplier No. AF7458
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Cytosolic Sulfotransferase 1C2/SULT1C2 Polyclonal specifically detects Cytosolic Sulfotransferase 1C2/SULT1C2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Cytosolic Sulfotransferase 1C2/SULT1C2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. O00338
Gene Alias cytosolic, 1C, member 1, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.1, EC 2.8.2.2, humSULTC2, ST1C1, ST1C2, Sulfotransferase 1C1, sulfotransferase family, cytosolic, 1C, member 2, SULT1C#1, SULT1C1, SULT1C2
Gene Symbols SULT1C2
Host Species Sheep
Immunogen E. coli-derived recombinant human Cytosolic Sulfotransferase 1C2/SULT1C2 Ala2-Leu296, predicted Accession # O00338
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6819
Test Specificity Detects human Cytosolic Sulfotransferase 1C2/SULT1C2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SULT1C4 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.