Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cytosolic Sulfotransferase 1E1/SULT1E1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5545SP
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 1E1/SULT1E1 Polyclonal specifically detects Cytosolic Sulfotransferase 1E1/SULT1E1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Cytosolic Sulfotransferase 1E1/SULT1E1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.8.2, EST-1, ESTMGC34459, estrone sulfotransferase, ST1E1, STE, STEestrogen sulfotransferase, Sulfotransferase 1E1, sulfotransferase family 1E, estrogen-preferring, member 1, Sulfotransferase, estrogen-preferringEC 2.8.2.4, SULT1E1 | |
Goat | |
Affinity Purified | |
RUO | |
6783 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
P49888 | |
SULT1E1 | |
E. coli-derived recombinant human Sulfotransferase 1E1/SULT1E1 Asn2-Ile294 Accession # P49888 | |
25 μg | |
Primary | |
Detects human Sulfotransferase 1E1/SULT1E1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Sulfotransferase 1A1 is observed and less than 2% cross-reactivity with rhSulfotransferase 2A1 and rhSulfotransferase 4A1 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction