Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cytosolic Sulfotransferase 2B1/SULT2B1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6174SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6174SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6174SP Supplier R&D Systems Supplier No. MAB6174SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cytosolic Sulfotransferase 2B1/SULT2B1 Monoclonal specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Cytosolic Sulfotransferase 2B1/SULT2B1
Applications Immunohistochemistry
Classification Monoclonal
Clone 682516
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O00204
Gene Alias Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1, SULT2B1
Gene Symbols SULT2B1
Host Species Mouse
Immunogen E. coli-derived recombinant human Cytosolic Sulfotransferase 2B1/SULT2B1 Asp2-Ser365 Accession # O00204
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6820
Test Specificity Detects human Cytosolic Sulfotransferase 2B1/SULT2B1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) SULT1A1, 1A3, 1B1, 1C4, 1E1, 2A1, or 4A1 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.