Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cytosolic Sulfotransferase 4A1/SULT4A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5826SP
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 4A1/SULT4A1 Monoclonal specifically detects Cytosolic Sulfotransferase 4A1/SULT4A1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Cytosolic Sulfotransferase 4A1/SULT4A1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
Q9BR01 | |
SULT4A1 | |
E. coli-derived recombinant human Cytosolic Sulfotransferase 4A1/SULT4A1 Ala2-Lys284 Accession # Q9BR01 | |
25 μg | |
Primary | |
Detects human Cytosolic Sulfotransferase 4A1/SULT4A1 in direct ELISAs and Western blots. No cross-reactivity with recombinant human (rh) SULT1A1, rhSULT1A3, rhSULT1B1, rhSULT1E1, rhSULT2A1, or rhSULT2B1 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
634630 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Brain sulfotransferase-like protein, BRSTL1, BR-STL-1, EC 2.8.2.-, hBR-STL, hBR-STL-1MGC40032, nervous system cytosolic sulfotransferase, Nervous system sulfotransferase, NST, ST4A1, sulfotransferase 4A1, sulfotransferase family 4A, member 1, sulfotransferase-related protein, SULT4A1, SULTX3, SULTX3DJ388M5.3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
25830 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction