Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DAN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$176.44 - $650.95
Specifications
| Antigen | DAN/NBL1 |
|---|---|
| Clone | 161502 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
Description
DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| DAN/NBL1 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN | |
| NBL1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 161502 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P41271 | |
| 4681 | |
| Mouse myeloma cell line NS0-derived recombinant human DAN Ala17-Asp180 Accession # P41271 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human DAN in ELISAs and Western blots. In ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) DAN, recombinant chicken Caronte, recombinant human Cerberus, or rmGremlin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title