Learn More
Human DAP10/HCST Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | DAP10/HCST |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 982129 |
| Conjugate | Alexa Fluor 350 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | Q9UBK5 |
| Gene Alias | DAP10PIK3AP, DKFZP586C1522, HCST, hematopoietic cell signal transducer, KAP10, KAP10DNAX-activation protein 10, kinase assoc pro of ∼10kDa, kinase assoc protein, Membrane protein DAP10, phosphoinositide-3-kinase adaptor protein, PIK3AP, Transmembrane adapter protein KAP10 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.