Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DARC Alexa Fluor 405-conjugated Antibody, R&D Systems™

Catalog No. FAB4139V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4139V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4139V100 Supplier R&D Systems Supplier No. FAB4139V100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DARC Monoclonal specifically detects DARC in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DARC
Applications Flow Cytometry
Classification Monoclonal
Clone 358307
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q16570
Gene Alias ACKR1, CCBP1, CD234, CD234 antigen, Dfy, Duffy Antigen, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy Antigen, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPD, GPDWBCQ1, GpFy, Plasmodium vivax receptor
Gene Symbols DARC
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human DARC Met1-Ser336 Accession # Q16570
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2532
Test Specificity Detects human DARC. Stains human DARC transfected cells but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.