Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DARC PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4139P
This item is not returnable.
View return policy
Description
DARC Monoclonal specifically detects DARC in Human samples. It is validated for Flow Cytometry.Specifications
DARC | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ACKR1, CCBP1, CD234, CD234 antigen, Dfy, Duffy Antigen, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy Antigen, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPD, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2532 | |
Human | |
Purified |
Flow Cytometry | |
358307 | |
Flow Cytometry 10 uL/10^6 cells | |
Q16570 | |
DARC | |
HEK293 human embryonic kidney cell line transfected with human DARC Met1-Ser336 Accession # Q16570 | |
100 Tests | |
Primary | |
Detects human DARC. Stains human DARC transfected cells but not irrelevant transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction