Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DC-SIGN/CD209 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
$186.30 - $374.00
Specifications
| Antigen | DC-SIGN/CD209 |
|---|---|
| Clone | 120507 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB161F025
![]() |
R&D Systems
FAB161F025 |
25 Tests |
Each for $186.30
|
|
|||||
FAB161F100
![]() |
R&D Systems
FAB161F100 |
100 Tests |
Each for $374.00
|
|
|||||
Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Flow Cytometry.Specifications
| DC-SIGN/CD209 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 30835 | |
| NIH-3T3 mouse embryonic fibroblast cell line transfected with human DC-SIGN/CD209 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 120507 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| Human | |
| CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 | |
| CD209 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human DC-SIGN/CD209 on transfected NIH/3T3 cells and on monocyte derived dendritic cells. Does not react with parental mouse cells or irrelevant transfectants, such as human DC-SIGN2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title