Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DC-STAMP APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB7824A
This item is not returnable.
View return policy
Description
DC-STAMP/TM7SF4 Monoclonal specifically detects DC-STAMP/TM7SF4 in Human samples. It is validated for Flow Cytometry.Specifications
DC-STAMP/TM7SF4 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
DCSTAMP, dendritic cell-specific transmembrane protein, FIND, IL-4-induced Protein, TM7SF4, transmembrane 7 superfamily member 4 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
81501 | |
Human | |
Purified |
Flow Cytometry | |
788524 | |
Flow Cytometry 10 uL/10^6 cells | |
Q9H295 | |
DCSTAMP | |
E. coli-derived recombinant human DC-STAMP Asp314-Thr376 Accession # Q9H295 | |
100 Tests | |
Primary | |
Detects human DC-STAMP in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse DC-STAMP is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction