Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DCC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5884SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5884SP 25 μg
MAB5884 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5884SP Supplier R&D Systems Supplier No. MAB5884SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DCC Monoclonal specifically detects DCC in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen DCC
Applications Immunohistochemistry
Classification Monoclonal
Clone 531505
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P43146
Gene Alias Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC
Gene Symbols DCC
Host Species Mouse
Immunogen E. coli-derived recombinant human DCC Ser1323-Phe1447 Accession # P43146
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1630
Test Specificity Detects human DCC in direct ELISAs.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.