Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DCC Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5884
This item is not returnable.
View return policy
Description
DCC Polyclonal specifically detects DCC in Human samples. It is validated for Western Blot.Specifications
DCC | |
Polyclonal | |
Unconjugated | |
P43146 | |
DCC | |
E. coli-derived recombinant human DCC Ser1323-Phe1447 Accession # P43146 | |
100 μg | |
Primary | |
Detects human DCC in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ICAM-1, rhICAM-2, rhICAM-3, rhMadCAM-1, rhPECAM-1, rhVCAM-1, and rhCEACAM-1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Colorectal cancer suppressor, CRC18, CRCR1, deleted in colorectal cancer protein, deleted in colorectal carcinoma, IGDCC1colorectal tumor suppressor, Immunoglobulin superfamily DCC subclass member 1, immunoglobulin superfamily, DCC subclass, member 1, netrin receptor DCC, Tumor suppressor protein DCC | |
Sheep | |
Affinity Purified | |
RUO | |
1630 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction