Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DCIR/CLEC4A Alexa Fluor™ 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1748S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1748S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1748S100 Supplier R&D Systems Supplier No. FAB1748S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

DCIR/CLEC4A Monoclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen DCIR/CLEC4A
Applications Flow Cytometry
Classification Monoclonal
Clone 216110
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q9UMR7
Gene Alias CD367, CLEC4A, CLECSF6, CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor, LLIR
Gene Symbols CLEC4A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human DCIR isoform 1 Gln70-Leu237 Accession # Q9UMR7
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50856
Test Specificity Detects human DCIR in direct ELISAs and Western blots.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.