Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DCIR/CLEC4A Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF1748
This item is not returnable.
View return policy
Description
DCIR/CLEC4A Polyclonal specifically detects DCIR/CLEC4A in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
DCIR/CLEC4A | |
Polyclonal | |
Biotin | |
Q9UMR7 | |
CLEC4A | |
Mouse myeloma cell line NS0-derived recombinant human DCIR/CLEC4A isoform 1 Gln70-Leu237 Accession # Q9UMR7 | |
50 μg | |
Primary | |
Detects human DCIR/CLEC4A in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
CD367, CLEC4A, CLECSF6, CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor, LLIR | |
Goat | |
Affinity Purified | |
RUO | |
50856 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction