Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human DCIR/CLEC4A ELISA Kit (Colorimetric)
SDP

Catalog No. NB231481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB231481 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB231481 Supplier Novus Biologicals Supplier No. NBP280344

Specifications

Assay Range 78.125-5000 pg/mL
Assay Sensitivity 13.18 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Lysates
Specificity Recognizes both recombinant and natural Human DCIR/CLEC4A.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 50856
Gene Symbol CLEC4A
Quantity 1 Kit
Research Discipline Immunology, Innate Immunity
Storage Requirements Storage is content dependent.
Synonym CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.