Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DCLK1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7138SP
This item is not returnable.
View return policy
Description
DCLK1 Polyclonal specifically detects DCLK1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DCLK1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CL1, CLICK1, DCAMKL1, DCAMKL1CLICK1, DCDC3A, DCDC3ACL1, DCLK, doublecortin and CaM kinase-like 1, Doublecortin domain-containing protein 3A, doublecortin-like kinase 1Doublecortin-like and CAM kinase-like 1, EC 2.7.11, KIAA0369EC 2.7.11.1, serine/threonine-protein kinase DCLK1 | |
Sheep | |
Affinity Purified | |
RUO | |
9201 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
NP_004725 | |
DCLK1 | |
E. coli-derived recombinant human DCLK1 Isoform 1 Lys621-Met729 Accession # NP_004725 | |
25 μg | |
Primary | |
Detects human DCLK1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction