Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human DcR3/TNFRSF6B Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF142
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF142 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF142 Supplier R&D Systems Supplier No. AF142
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DcR3/TNFRSF6B Polyclonal specifically detects DcR3/TNFRSF6B in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.

Specifications

Antigen DcR3/TNFRSF6B
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.1-0.5 ug/mL
Gene Accession No. O95407
Gene Alias DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy
Gene Symbols TNFRSF6B
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human DcR3/TNFRSF6B Val24-His300 Accession # O95407
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8771
Test Specificity Detects human DcR3/TNFRSF6B in ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) TRAIL R1, rhTRAIL R2, rhTRAIL R3, and rhTRAIL R4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.