Learn More
Human DDR2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | DDR2 |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Clone | 290804 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q16832 |
| Gene Alias | CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKT, TKTMIG20a, Trk3, Tyro-10, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.