Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Decorin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB143SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB143SP 25 μg
MAB143100 100 μg
MAB143500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB143SP Supplier R&D Systems Supplier No. MAB143SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Decorin Monoclonal specifically detects Decorin in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Decorin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 115402
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Simple Western 10 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_598013.1
Gene Alias Bone proteoglycan II, CSCD, DCN, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-II, PGS2, PG-S2, proteoglycan core protein, SLRR1B, SLRR1BPGS2, small leucine-rich protein 1B
Gene Symbols DCN
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Decorin Gly17-Lys172 Accession # NP_598013.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1634
Test Specificity Detects human Decorin in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse Decorin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.