Learn More
Human Dectin-1/CLEC7A PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 9 publications
Supplier:  R&D Systems FAB1859P025
 View more versions of this product
Catalog No. FAB1859P025
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Dectin-1/CLEC7A Monoclonal specifically detects Dectin-1/CLEC7A in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Dectin-1/CLEC7A
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
64581
Human
Purified
Flow Cytometry
259931
Flow Cytometry 10 uL/10^6 cells
NP_072092
CLEC7A
Mouse myeloma cell line NS0-derived recombinant human Dectin-1/CLEC7A Thr66-Met201 Accession # NP_072092
25 Tests
Primary
Detects human Dectin-1/CLEC7A in direct ELISAs. In directs ELISAs, less than 10% cross-reactivity with recombinant human (rh) DLEC and no cross-reactivity with recombinant mouse Dectin-1 or rhDectin-2 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.