Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Dectin-2/CLEC6A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB31141100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB31141100 100 μg
MAB31141SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB31141100 Supplier R&D Systems Supplier No. MAB31141100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Dectin-2/CLEC6A Monoclonal specifically detects Dectin-2/CLEC6A in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Dectin-2/CLEC6A
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 545925
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q6EIG7
Gene Alias CLEC4N, CLEC6A, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Dectin-2/CLEC6A, Thr46-Leu209, Accession # Q6EIG7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93978
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.