Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DEP-1/CD148 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Supplier: R&D Systems MAB1934SP
This item is not returnable.
View return policy
Description
DEP-1/CD148 Monoclonal specifically detects DEP-1/CD148 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF-ready.Specifications
DEP-1/CD148 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunoprecipitation 2.5 ug/500 ug cell lysate, CyTOF-ready | |
CD148, CD148 antigen, Density-enhanced phosphatase 1, DEP1, DEP-1, DEP1EC 3.1.3.48, HPTP eta, HPTPeta, HPTP-eta, human density enhanced phosphatase-1, protein tyrosine phosphatase, receptor type, J, protein tyrosine phosphatase, receptor type, J polypeptide, Protein-tyrosine phosphatase eta, Protein-tyrosine phosphatase receptor type J, PTPRJ, receptor-type tyrosine-protein phosphatase eta, R-PTP-eta, R-PTP-J, SCC1, susceptibility to colon cancer 1, mouse, homolog of | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5795 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Flow Cytometry, Immunoprecipitation, CyTOF | |
143-41 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
PTPRJ | |
Phytohemagglutinin-stimulated human peripheral blood mononuclear cells | |
25 μg | |
Primary | |
Detects human DEP-1/CD148 in Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction