Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human DEPTOR/DEPDC6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7255
This item is not returnable.
View return policy
Description
DEPTOR/DEPDC6 Polyclonal specifically detects DEPTOR/DEPDC6 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DEPTOR/DEPDC6 | |
Polyclonal | |
Unconjugated | |
Q8TB45 | |
DEPTOR | |
E. coli-derived recombinant human DEPTOR/DEPDC6 Ala31-Gly158 Accession # Q8TB45 | |
100 μg | |
Primary | |
Detects recombinant human DEPTOR/DEPDC6 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
DEP domain containing 6, DEP domain containing mTOR interacting protein, DEP domain containing MTOR-interacting protein, DEP domain-containing mTOR-interacting protein, DEP domain-containing protein 6, DEP.6, DEPDC6, DEPDC6FLJ12341, DKFZp564B1778, FLJ12428, FLJ13854 | |
Sheep | |
Affinity Purified | |
RUO | |
64798 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction