Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Dermatopontin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4629SP
This item is not returnable.
View return policy
Description
Dermatopontin Polyclonal specifically detects Dermatopontin in Human samples. It is validated for Western Blot.Specifications
Dermatopontin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
dermatopontin, DPT, Eq1, TRAMP, Tyrosine-rich acidic matrix protein | |
Sheep | |
Affinity Purified | |
RUO | |
1805 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AAH33736 | |
DPT | |
Mouse myeloma cell line NS0-derived recombinant human Dermatopontin Gln19-Val201 Accession # AAH33736 | |
25 μg | |
Primary | |
Detects human Dermatopontin in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse Dermatopontin is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction