Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Desmocollin-1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$538.00
Specifications
Antigen | Desmocollin-1 |
---|---|
Clone | 2906A |
Dilution | Immunohistochemistry 3-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB4955100
![]() |
R&D Systems
MAB4955100 |
100 μg |
Each of 1 for $538.00
|
|
|||||
Description
Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Human samples. It is validated for ImmunohistochemistrySpecifications
Desmocollin-1 | |
Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, Desmocollin1, desmosomal glycoprotein 2/3, DG2/DG3, DSC1 | |
Mouse myeloma cell line, NS0-derived human Desmocollin-1, Arg135-Asn686, Accession # Q08554 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2906A | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q08554 | |
1823 | |
IgG | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title