Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Desmocollin-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4688
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4688 100 μg
AF4688SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4688 Supplier R&D Systems Supplier No. AF4688
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Desmocollin-2 Polyclonal specifically detects Desmocollin-2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Desmocollin-2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q02487
Gene Alias ARVD11, Cadherin family member 2, CDHF2, CDHF2DGII/III, desmocollin 2, desmocollin-2, Desmocollin2, Desmocollin-3, Desmosomal glycoprotein II, desmosomal glycoprotein II/III, Desmosomal glycoprotein III, DG2, DSC2, DSC3DKFZp686I11137
Gene Symbols DSC2
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Desmocollin-2 Arg136-Arg684 Accession # Q02487
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1824
Test Specificity Detects human Desmocollin-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) Desmocollin-1 and rhDesmocollin-3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.